2,6-Dimethoxyphenylboronic acid ≥97% (contains varying amounts of Anhydride)
Supplier: TCI America
D3861-25G
D3861-5G
TCD3861-25GEA
308.1
CAD
TCD3861-25G
TCD3861-5G
2,6-Dimethoxyphenylboronic acid ≥97% (contains varying amounts of Anhydride)
2,6-Dimethoxyphenylboronic acid
CAS Number: 23112-96-1
MDL Number: MFCD01318987
Molecular Formula: C8H11BO4
Molecular Weight: 181.98
Form: Crystal
Color: White
Melting point (°C): 113
Other Product Names / Synonyms: 2,6-Dimethoxybenzeneboronic Acid
Formula:
C₈H₁₁BO₄ MW: 181.98 g/mol Melting Pt: 113 °C |
MDL Number:
MFCD01318987 CAS Number: 23112-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...