4,4-Dimethyl-2-cyclohexen-1-one ≥96.0%
Supplier: TCI America
Synonyms:
4,4-Dimethylcyclohex-2-enone
D3852-25G
D3852-5G
TCD3852-25GEA
828.08
CAD
TCD3852-25G
TCD3852-5G
4,4-Dimethyl-2-cyclohexen-1-one ≥96.0%
4,4-Dimethyl-2-cyclohexen-1-one
CAS Number: 1073-13-8
MDL Number: MFCD00009695
Molecular Formula: C8H12O
Molecular Weight: 124.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 74
Flash Point (°C): 64
Specific Gravity (20/20): 0.94
Lambda max.: 318 nm (EtOH)
Formula:
C₈H₁₂O Boiling Pt: 74 °C Density: 0.94 (20 °C) Flash Pt: 64 °C |
MDL Number:
MFCD00009695 CAS Number: 1073-13-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...