N-(Diphenylmethylene)glycine ethyl ester ≥97.0%

Supplier: TCI America

Synonyms: Ethyl (diphenylmethylenamino)acetate, Ethyl 2-((diphenylmethylene)amino)acetate, Ethyl-2-((diphenylmethylene)amino)acetate, Ethyl N-(diphenylmethylene)glycinate

D3833-25G D3833-5G
TCD3833-25GEA 259.12 CAD
TCD3833-25G TCD3833-5G
N-(Diphenylmethylene)glycine ethyl ester ≥97.0%
N-(Diphenylmethylene)glycine ethyl ester

CAS Number: 69555-14-2
MDL Number: MFCD00010590
Molecular Formula: C17H17NO2
Molecular Weight: 267.33
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 195
Melting point (°C): 52
Flash Point (°C): 110
Lambda max.: 247 nm (Hexane)

Formula: C₁₇H₁₇NO₂
MW: 267.32 g/mol
Boiling Pt: 195 °C
Melting Pt: 52 °C
Flash Pt: 110 °C
MDL Number: MFCD00010590
CAS Number: 69555-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d0737114feb2b6a71dfcef421614f63fb2a2410&vid=8d0737114feb2b6a71dfcef421614f63fb2a2410&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR