Diethyl (hydroxymethyl)phosphonate ≥97.0%
Supplier: TCI America
D3813-100G
D3813-25G
TCD3813-100GEA
363.16
CAD
TCD3813-100G
TCD3813-25G
Diethyl (hydroxymethyl)phosphonate ≥97.0%
Diethyl (hydroxymethyl)phosphonate
CAS Number: 3084-40-0
MDL Number: MFCD00014418
Molecular Formula: C5H13O4P
Molecular Weight: 168.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 113
Specific Gravity (20/20): 1.17
Storage Temperature: 0-10°C
Other Product Names / Synonyms: (Hydroxymethyl)phosphonic Acid Diethyl Ester
Formula:
C₅H₁₃O₄P Boiling Pt: 105 °C Density: 1.17 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00014418 CAS Number: 3084-40-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...