2,6-Dichloro-4-(trifluoromethoxy)aniline ≥98.0%
Supplier: TCI America
D3802-5G
D3802-25G
TCD3802-5GEA
122.29
CAD
TCD3802-5G
TCD3802-25G
2,6-Dichloro-4-(trifluoromethoxy)aniline ≥98.0%
2,6-Dichloro-4-(trifluoromethoxy)aniline
CAS Number: 99479-66-0
MDL Number: MFCD00190128
Molecular Formula: C7H4Cl2F3NO
Molecular Weight: 246.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 33
Flash Point (°C): 113
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Amino-1,3-dichloro-5-(trifluoromethoxy)benzene
Formula:
C₇H₄Cl₂F₃NO Melting Pt: 33 °C Flash Pt: 113 °C |
MDL Number:
MFCD00190128 CAS Number: 99479-66-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...