2,6-Dichlorobenzothiazole ≥98.0%

Supplier: TCI America

D3801-1G D3801-5G
TCD3801-1GEA 109.02 CAD
TCD3801-1G TCD3801-5G
2,6-Dichlorobenzothiazole ≥98.0%
2,6-Dichlorobenzothiazole

CAS Number: 3622-23-9
MDL Number: MFCD00044101
Molecular Formula: C7H3Cl2NS
Molecular Weight: 204.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 98

Formula: C₇H₃Cl₂NS
Melting Pt: 98 °C
MDL Number: MFCD00044101
CAS Number: 3622-23-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55bd96a775c545604329199637f3a5334e8040df&vid=55bd96a775c545604329199637f3a5334e8040df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR