3,5-Dichlorobenzyl alcohol ≥98.0%
Supplier: TCI America
D3791-25G
D3791-5G
TCD3791-25GEA
186.76
CAD
TCD3791-25G
TCD3791-5G
3,5-Dichlorobenzyl alcohol ≥98.0%
3,5-Dichlorobenzyl alcohol
CAS Number: 60211-57-6
MDL Number: MFCD00004634
Molecular Formula: C7H6Cl2O
Molecular Weight: 177.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 79
Formula:
C₇H₆Cl₂O Melting Pt: 79 °C |
MDL Number:
MFCD00004634 CAS Number: 60211-57-6 UN: 2250 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...