EDTA disodium salt dihydrate ≥99.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

D3789-25G D3789-5G
TCD3789-25GEA 135.88 CAD
TCD3789-25G TCD3789-5G
EDTA disodium salt dihydrate ≥99.0% (by titrimetric analysis) for biochemical research
EDTA disodium salt dihydrate

[for Biochemical Research]
CAS Number: 6381-92-6
MDL Number: MFCD00150037
Molecular Formula: C10H16N2O8
Molecular Weight: 336.21
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 252


Product Categories: Bioscience, Protease Inhibitors (for Biochemical Research)
Other Product Names / Synonyms: Ethylenediaminetetraacetic Acid Disodium Salt, EDTA 2Na, ETA

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255
MDL Number: MFCD00150037
CAS Number: 6381-92-6
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=836ce9c82f4b93caec6fda27a8c2629cd6a4f34d&vid=86a6eac252e8b270fc2c3f969f91d125ebd754e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR