2,4-Dichloropyridine ≥98.0%

Supplier: TCI America

D3784-25G D3784-5G
TCD3784-25GEA 205.4 CAD
TCD3784-25G TCD3784-5G
2,4-Dichloropyridine ≥98.0%
2,4-Dichloropyridine

CAS Number: 26452-80-2
MDL Number: MFCD00955618
Molecular Formula: C5H3Cl2N
Molecular Weight: 147.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 190
Flash Point (°C): 89
Specific Gravity (20/20): 1.37

Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 190 °C
Density: 1.37 (20 °C)
Flash Pt: 89 °C
MDL Number: MFCD00955618
CAS Number: 26452-80-2
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e72f5723ab3a70a7cf8093bfad3b72efad69df0e&vid=e72f5723ab3a70a7cf8093bfad3b72efad69df0e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR