3,4-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
D3783-25G
D3783-5G
TCD3783-25GEA
249.64
CAD
TCD3783-25G
TCD3783-5G
3,4-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
3,4-Dichlorophenylboronic acid
CAS Number: 151169-75-4
MDL Number: MFCD01074646
Molecular Formula: C6H5BCl2O2
Molecular Weight: 190.81
Form: Crystal
Color: White
Melting point (°C): 285
Other Product Names / Synonyms: 3,4-Dichlorobenzeneboronic Acid
Formula:
C₆H₅BCl₂O₂ Melting Pt: 285 °C |
MDL Number:
MFCD01074646 CAS Number: 151169-75-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...