2,2'-Dithienyl disulfide ≥97.0% (by HPLC)

Supplier: TCI America

D3774-1G D3774-5G
TCD3774-1GEA 58.46 CAD
TCD3774-1G TCD3774-5G
2,2'-Dithienyl disulfide ≥97.0% (by HPLC)
2,2'-Dithienyl disulfide

CAS Number: 6911-51-9
MDL Number: MFCD00066333
Molecular Formula: C8H6S4
Molecular Weight: 230.38
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Boiling point (°C): 132
Melting point (°C): 58


Other Product Names / Synonyms: Di(2-thienyl) Disulfide

Formula: C₈H₆S₄
Boiling Pt: 132 °C
Melting Pt: 58 °C
MDL Number: MFCD00066333
CAS Number: 6911-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb39e95d8c4302db10d2e8165b9719a4c4442bfc&vid=eb39e95d8c4302db10d2e8165b9719a4c4442bfc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR