(3,4-Difluorophenyl)acetic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

D3771-1G D3771-5G
TCD3771-1GEA 48.98 CAD
TCD3771-1G TCD3771-5G
(3,4-Difluorophenyl)acetic acid ≥97.0% (by GC, titration analysis)
(3,4-Difluorophenyl)acetic acid

CAS Number: 658-93-5
MDL Number: MFCD00010002
Molecular Formula: C8H6F2O2
Molecular Weight: 172.13
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 50

Formula: (C₆H₃F₂)CH₂COOH
Melting Pt: 50 °C
MDL Number: MFCD00010002
CAS Number: 658-93-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=009a297fe0d89b1cb5f46f1caff366f85c13ee1e&vid=009a297fe0d89b1cb5f46f1caff366f85c13ee1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR