2,4-Dichloro-5-nitropyrimidine ≥98.0%

Supplier: TCI America

D3721-25G D3721-5G
TCD3721-25GEA 268.6 CAD
TCD3721-25G TCD3721-5G
2,4-Dichloro-5-nitropyrimidine ≥98.0%
2,4-Dichloro-5-nitropyrimidine

CAS Number: 49845-33-2
MDL Number: MFCD00127867
Molecular Formula: C4HCl2N3O2
Molecular Weight: 193.97
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 139
Flash Point (°C): 110
Freezing point (°C): 28
Storage Temperature: 0-10°C

Formula: C₄HCl₂N₃O₂
MW: 193.98 g/mol
Boiling Pt: 139 °C
Flash Pt: 110 °C
MDL Number: MFCD00127867
CAS Number: 49845-33-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e13203cde318768265384533b3246f59f2058ca7&vid=e13203cde318768265384533b3246f59f2058ca7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR