Diphenylphosphinic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D3627-25G D3627-5G
TCD3627-25GEA 178.54 CAD
TCD3627-25G TCD3627-5G
Diphenylphosphinic acid ≥98.0% (by HPLC, titration analysis)
Diphenylphosphinic acid

CAS Number: 1707-03-5
MDL Number: MFCD00002132
Molecular Formula: C12H11O2P
Molecular Weight: 218.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 195

Formula: C₁₂H₁₁O₂P
Melting Pt: 195 °C
MDL Number: MFCD00002132
CAS Number: 1707-03-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=520fe85103f5f028adc108b240844345ae886dd8&vid=520fe85103f5f028adc108b240844345ae886dd8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR