1,2-Difluoro-4,5-dimethoxybenzene ≥95.0%
Supplier: TCI America
D3626-1G
D3626-5G
TCD3626-1GEA
74.26
CAD
TCD3626-1G
TCD3626-5G
1,2-Difluoro-4,5-dimethoxybenzene ≥95.0%
1,2-Difluoro-4,5-dimethoxybenzene
CAS Number: 203059-80-7
MDL Number: MFCD00070789
Molecular Formula: C8H8F2O2
Molecular Weight: 174.15
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 43
Flash Point (°C): 92
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4,5-Difluoroveratrole
Formula:
C₈H₈F₂O₂ MW: 174.15 g/mol Melting Pt: 43 °C Flash Pt: 92 °C |
MDL Number:
MFCD00070789 CAS Number: 203059-80-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...