Dichloro(1,5-cyclooctadiene)platinum(II)

Supplier: TCI America

Synonyms: 1,5-Cyclooctadieneplatinum(II) Dichloride

D3592-1G D3592-200MG
TCD3592-1GEA 534.04 CAD
TCD3592-1G TCD3592-200MG
Dichloro(1,5-cyclooctadiene)platinum(II)
Dichloro(1,5-cyclooctadiene)platinum(II)

CAS Number: 12080-32-9
MDL Number: MFCD00012413
Molecular Formula: C8H12Cl2Pt
Molecular Weight: 374.17
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 285


Other Product Names / Synonyms: 1,5-Cyclooctadieneplatinum(II) Dichloride

Formula: C₈H₁₂Cl₂Pt
MW: 374.17 g/mol
Melting Pt: 285 °C
MDL Number: MFCD00012413
CAS Number: 12080-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6613c78b442b1c569f81b3e2ec267cbb70bd952&vid=e6613c78b442b1c569f81b3e2ec267cbb70bd952&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR