Dimethyl octafluoroadipate ≥97.0%

Supplier: TCI America

D3590-5G D3590-1G
TCD3590-5GEA 48.98 CAD
TCD3590-5G TCD3590-1G
Dimethyl octafluoroadipate ≥97.0%
Dimethyl octafluoroadipate

CAS Number: 3107-98-0
MDL Number: MFCD00042267
Molecular Formula: C8H6F8O4
Molecular Weight: 318.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Specific Gravity (20/20): 1.55


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Dimethyl Perfluoroadipate, Dimethyl Octafluorohexanedioate, Octafluoroadipic Acid Dimethyl Ester, Perfluoroadipic Acid Dimethyl Ester, Octafluorohexanedioic Acid Dimethyl Ester

Formula: C₈H₆F₈O₄
Boiling Pt: 123 °C
Density: 1.55 (20 °C)
MDL Number: MFCD00042267
CAS Number: 3107-98-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d261a1b054caa113443a76a4b9e7c0ec4a61aefd&vid=d261a1b054caa113443a76a4b9e7c0ec4a61aefd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR