N,N-Dimethylglycine ≥98.0%

Supplier: TCI America

Synonyms: 2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid

D3585-25G D3585-5G
TCD3585-25GEA 254 CAD
TCD3585-25G TCD3585-5G
N,N-Dimethylglycine ≥98.0%
N,N-Dimethylglycine

CAS Number: 1118-68-9
MDL Number: MFCD00004283
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 180

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: 180 °C
MDL Number: MFCD00004283
CAS Number: 1118-68-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=024a2949a1090a30d2991e2396344e08e3b4304e&vid=024a2949a1090a30d2991e2396344e08e3b4304e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR