2,6-Dibromo-p-toluidine ≥98.0%
Supplier: TCI America
Synonyms:
4-Amino-3,5-dibromotoluene, 2,6-Dibromo-4-methylaniline
D3577-25G
D3577-5G
TCD3577-25GEA
303.36
CAD
TCD3577-25G
TCD3577-5G
2,6-Dibromo-p-toluidine ≥98.0%
2,6-Dibromo-p-toluidine
CAS Number: 6968-24-7
MDL Number: MFCD00007641
Molecular Formula: C7H7Br2N
Molecular Weight: 264.95
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 74
Other Product Names / Synonyms: 3,5-Dibromo-4-aminotoluene, 2,6-Dibromo-p-toluidine
Formula:
C₇H₇Br₂N MW: 264.95 Boiling Pt: 296 Melting Pt: 79 |
MDL Number:
MFCD00007641 CAS Number: 6968-24-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...