3,4-Difluorobenzophenone ≥98.0%
Supplier: TCI America
D3576-5G
D3576-25G
TCD3576-5GEA
105.86
CAD
TCD3576-5G
TCD3576-25G
3,4-Difluorobenzophenone ≥98.0%
3,4-Difluorobenzophenone
CAS Number: 85118-07-6
MDL Number: MFCD00009892
Molecular Formula: C13H8F2O
Molecular Weight: 218.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 55
Flash Point (°C): 113
Formula:
C₁₃H₈F₂O Melting Pt: 55 °C Flash Pt: 113 °C |
MDL Number:
MFCD00009892 CAS Number: 85118-07-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...