1,6-Dibromoperfluorohexane ≥98.0%
Supplier: TCI America
Synonyms:
Perfluoro(1,6-dibromohexane)
D3572-1G
D3572-5G
TCD3572-1GEA
52.14
CAD
TCD3572-1G
TCD3572-5G
1,6-Dibromoperfluorohexane ≥98.0%
1,6-Dibromoperfluorohexane
CAS Number: 918-22-9
MDL Number: MFCD00153110
Molecular Formula: C6Br2F12
Molecular Weight: 459.85
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Specific Gravity (20/20): 2.10
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,6-Dibromoperfluorohexane
Formula:
C₆Br₂F₁₂ MW: 459.87 Boiling Pt: 150 °C Density: 2.10 (20 °C) |
MDL Number:
MFCD00153110 CAS Number: 918-22-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...