5'-O-(4,4'-Dimethoxytrityl)thymidine ≥98.0%
Supplier: TCI America
Synonyms:
DMT-T, 1-((2R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxytetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, 5'-O-Dimethoxytrityl-deoxythymidine
CAS Number: 40615-39-2
MDL Number: MFCD00010113
Molecular Formula: C31H32N2O7
Molecular Weight: 544.60
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 151
Specific rotation [a]20/D: 7 deg (C=1, MeOH)
Storage Temperature: 0-10°C
Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: DMT-T
Formula:
C₃₁H₃₂N₂O₇ Melting Pt: 151 °C |
MDL Number:
MFCD00010113 CAS Number: 40615-39-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...