2',6'-Difluoroacetophenone ≥98.0%

Supplier: TCI America

Synonyms: 1-(2,6-Difluorophenyl)ethan-1-one, 1-(2,6-Difluorophenyl)ethanone

D3559-25G D3559-5G
TCD3559-25GEA 413.96 CAD
TCD3559-25G TCD3559-5G
2',6'-Difluoroacetophenone ≥98.0%
2',6'-Difluoroacetophenone

CAS Number: 13670-99-0
MDL Number: MFCD00000328
Molecular Formula: C8H6F2O
Molecular Weight: 156.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 79
Flash Point (°C): 76
Specific Gravity (20/20): 1.24

Formula: C₈H₆F₂O
MW: 156.13 g/mol
Boiling Pt: 79 °C
Density: 1.24 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00000328
CAS Number: 13670-99-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87f2b38cd8d6be85926fd42e19859c5c435a9bce&vid=87f2b38cd8d6be85926fd42e19859c5c435a9bce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR