2,7-Dibromo-9-fluorenone ≥98.0%

Supplier: TCI America

D3557-5G D3557-25G
TCD3557-5GEA 42.66 CAD
TCD3557-5G TCD3557-25G
2,7-Dibromo-9-fluorenone ≥98.0%
2,7-Dibromo-9-fluorenone

CAS Number: 14348-75-5
MDL Number: MFCD00010790
Molecular Formula: C13H6Br2O
Molecular Weight: 338.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 204

Formula: C₁₃H₆Br₂O
Melting Pt: 204 °C
MDL Number: MFCD00010790
CAS Number: 14348-75-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cca7d21308ee3dc37c4b5281c1b7465fa8d4d33&vid=4cca7d21308ee3dc37c4b5281c1b7465fa8d4d33&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR