4,5-Difluoro-2-nitrobenzoic acid ≥98.0%
Supplier: TCI America
D3555-25G
D3555-5G
TCD3555-25GEA
301.78
CAD
TCD3555-25G
TCD3555-5G
4,5-Difluoro-2-nitrobenzoic acid ≥98.0%
4,5-Difluoro-2-nitrobenzoic acid
CAS Number: 20372-63-8
MDL Number: MFCD00799521
Molecular Formula: C7H3F2NO4
Molecular Weight: 203.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 165
Formula:
C₇H₃F₂NO₄ Melting Pt: 165 °C |
MDL Number:
MFCD00799521 CAS Number: 20372-63-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...