2,6-Dibromo-4-fluorophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1,3-Dibromo-5-fluoro-2-hydroxybenzene
D3531-5G
D3531-25G
TCD3531-5GEA
86.9
CAD
TCD3531-5G
TCD3531-25G
2,6-Dibromo-4-fluorophenol ≥98.0% (by GC, titration analysis)
2,6-Dibromo-4-fluorophenol
CAS Number: 344-20-7
MDL Number: MFCD00002317
Molecular Formula: C6H3Br2FO
Molecular Weight: 269.90
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 56
Formula:
C₆H₃Br₂FO MW: 269.9 g/mol Melting Pt: 56 °C |
MDL Number:
MFCD00002317 CAS Number: 344-20-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...