2,3-Difluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Dihydroxy(2,3-difluorophenyl)borane, 2,3-Difluorobenzeneboronic acid, (2,3-Difluorophenyl)metaboric acid

D3523-25G D3523-5G
TCD3523-25GEA 193.39 CAD
TCD3523-25G TCD3523-5G
2,3-Difluorophenylboronic acid (contains varying amounts of Anhydride)
2,3-Difluorophenylboronic acid

CAS Number: 121219-16-7
MDL Number: MFCD01863170
Molecular Formula: C6H5BF2O2
Molecular Weight: 157.91
Form: Crystal
Color: White


Other Product Names / Synonyms: 2,3-Difluorobenzeneboronic Acid

Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
MDL Number: MFCD01863170
CAS Number: 121219-16-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8c7891890ec96d72cee821ab14cd2d63effa2ef&vid=f8c7891890ec96d72cee821ab14cd2d63effa2ef&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR