2,5-Dimethoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(2,5-Dimethoxyphenyl)metaboric acid, 2,5-Dimethoxybenzeneboronic acid, (2,5-Dimethoxyphenyl)boronic acid, Dihydroxy(2,5-dimethoxyphenyl)borane
D3522-1G
D3522-5G
TCD3522-1GEA
50.56
CAD
TCD3522-1G
TCD3522-5G
2,5-Dimethoxyphenylboronic acid (contains varying amounts of Anhydride)
2,5-Dimethoxyphenylboronic acid
CAS Number: 107099-99-0
MDL Number: MFCD01318181
Molecular Formula: C8H11BO4
Molecular Weight: 181.98
Form: Crystal
Color: White
Melting point (°C): 97
Other Product Names / Synonyms: 2,5-Dimethoxybenzeneboronic Acid
Formula:
C₈H₁₁BO₄ MW: 181.98 Melting Pt: 97 °C |
MDL Number:
MFCD01318181 CAS Number: 107099-99-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...