2,4-Dimethoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

D3521-5G D3521-1G
TCD3521-5GEA 109.02 CAD
TCD3521-5G TCD3521-1G
2,4-Dimethoxyphenylboronic acid (contains varying amounts of Anhydride)
2,4-Dimethoxyphenylboronic acid

CAS Number: 133730-34-4
MDL Number: MFCD01074590
Molecular Formula: C8H11BO4
Molecular Weight: 181.98
Form: Crystal
Color: White
Melting point (°C): 105


Other Product Names / Synonyms: 2,4-Dimethoxybenzeneboronic Acid

Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
Melting Pt: 105 °C
MDL Number: MFCD01074590
CAS Number: 133730-34-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8181efd617f47f44527e4d6b04340d815464d7c5&vid=8181efd617f47f44527e4d6b04340d815464d7c5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR