2,6-Dimethoxynaphthalene ≥98.0%

Supplier: TCI America

D3519-5G D3519-25G
TCD3519-5GEA 235.42 CAD
TCD3519-5G TCD3519-25G
2,6-Dimethoxynaphthalene ≥98.0%
2,6-Dimethoxynaphthalene

CAS Number: 5486-55-5
MDL Number: MFCD00004063
Molecular Formula: C12H12O2
Molecular Weight: 188.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 154

Formula: C₁₂H₁₂O₂
Melting Pt: 154 °C
MDL Number: MFCD00004063
CAS Number: 5486-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52060ffd0c4452025206bde137a46e72e92af860&vid=52060ffd0c4452025206bde137a46e72e92af860&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR