2,5-Dimethylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

D3517-1G D3517-5G D3517-25G
TCD3517-1GEA 33.18 CAD
TCD3517-1G TCD3517-5G TCD3517-25G
2,5-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
2,5-Dimethylphenylboronic acid

CAS Number: 85199-06-0
MDL Number: MFCD01863525
Molecular Formula: C8H11BO2
Molecular Weight: 149.98
Form: Crystal
Color: White
Melting point (°C): 192


Other Product Names / Synonyms: 2,5-Dimethylbenzeneboronic Acid, p-Xylene-2-boronic Acid

Formula: C₈H₁₁BO₂
Melting Pt: 192 °C
MDL Number: MFCD01863525
CAS Number: 85199-06-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a10d1dd16b4a78184c69e4be460ed0a36b32497e&vid=a10d1dd16b4a78184c69e4be460ed0a36b32497e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR