2,4-Dimethylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

D3514-1G D3514-5G
TCD3514-1GEA 33.18 CAD
TCD3514-1G TCD3514-5G
2,4-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
2,4-Dimethylphenylboronic acid

CAS Number: 55499-44-0
MDL Number: MFCD02683101
Molecular Formula: C8H11BO2
Molecular Weight: 149.98
Form: Crystal
Color: White


Other Product Names / Synonyms: 2,4-Dimethylbenzeneboronic Acid, m-Xylene-4-boronic Acid

Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
MDL Number: MFCD02683101
CAS Number: 55499-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=398b02c92bc10a29958924cd102d4cf7ece0a69b&vid=398b02c92bc10a29958924cd102d4cf7ece0a69b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR