3,4-Dimethoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

D3512-5G D3512-25G
TCD3512-5GEA 140.62 CAD
TCD3512-5G TCD3512-25G
3,4-Dimethoxyphenylboronic acid (contains varying amounts of Anhydride)
3,4-Dimethoxyphenylboronic acid

CAS Number: 122775-35-3
MDL Number: MFCD01074574
Molecular Formula: C8H11BO4
Molecular Weight: 181.98
Form: Crystal
Color: White
Melting point (°C): 250


Other Product Names / Synonyms: 3,4-Dimethoxybenzeneboronic Acid

Formula: C₈H₁₁BO₄
Melting Pt: 250 °C
MDL Number: MFCD01074574
CAS Number: 122775-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd0a4c1b20574979016c14bbd43afa7efda4e66d&vid=fd0a4c1b20574979016c14bbd43afa7efda4e66d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR