Tetrahydrothiophen-3-one ≥98.0%

Supplier: TCI America

Synonyms: Dihydrothiophen-3(2H)-one, 4,5-Dihydrothiophen-3(2H)-one, 4,5-Dihydro-3(2H)-thiophenone

D3511-25G D3511-5G
TCD3511-25GEA 86.9 CAD
TCD3511-25G TCD3511-5G
Tetrahydrothiophen-3-one ≥98.0%
Tetrahydrothiophen-3-one

CAS Number: 1003-04-9
MDL Number: MFCD00005412
Molecular Formula: C4H6OS
Molecular Weight: 102.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 78
Specific Gravity (20/20): 1.20


Other Product Names / Synonyms: Tetrahydrothiophen-3-one, 3-Thiophanone

Formula: C₄H₆OS
Boiling Pt: 175 °C
Density: 1.20 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00005412
CAS Number: 1003-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7ef71520507a16414e1ca2dbbaa5cf99f1cf8c9&vid=b7ef71520507a16414e1ca2dbbaa5cf99f1cf8c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR