3,5-Dimethylbenzaldehyde ≥96.0%
Supplier: TCI America
D3498-5G
D3498-25G
TCD3498-5GEA
102.7
CAD
TCD3498-5G
TCD3498-25G
3,5-Dimethylbenzaldehyde ≥96.0%
3,5-Dimethylbenzaldehyde
CAS Number: 5779-95-3
MDL Number: MFCD00082777
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Melting point (°C): 9
Flash Point (°C): 99
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: m-Xylene-5-carboxaldehyde
Formula:
C₉H₁₀O Boiling Pt: 108 °C Melting Pt: 9 °C Density: 1.00 (20 °C) Flash Pt: 99 °C |
MDL Number:
MFCD00082777 CAS Number: 5779-95-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...