2,4-Dibromo-1-fluorobenzene ≥97.0%
Supplier: TCI America
Synonyms:
1,3-Dibromo-4-fluorobenzene, 1-Fluoro-2,4-dibromobenzene
D3489-25G
D3489-5G
TCD3489-25GEA
241.27
CAD
TCD3489-25G
TCD3489-5G
2,4-Dibromo-1-fluorobenzene ≥97.0%
2,4-Dibromo-1-fluorobenzene
CAS Number: 1435-53-6
MDL Number: MFCD00000283
Molecular Formula: C6H3Br2F
Molecular Weight: 253.90
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 92
Specific Gravity (20/20): 2.05
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,3-Dibromo-4-fluorobenzene
Formula:
C₆H₃Br₂F MW: 253.9 g/mol Boiling Pt: 105 °C Density: 2.05 (20 °C) Flash Pt: 92 °C |
MDL Number:
MFCD00000283 CAS Number: 1435-53-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...