2,3-Dimethylmaleic anhydride ≥98.0%

Supplier: TCI America

Synonyms: 3,4-Dimethylfuran-2,5-dione

D3473-5G D3473-25G
TCD3473-5GEA 136.61 CAD
TCD3473-5G TCD3473-25G
2,3-Dimethylmaleic anhydride ≥98.0%
2,3-Dimethylmaleic anhydride

CAS Number: 766-39-2
MDL Number: MFCD00005523
Molecular Formula: C6H6O3
Molecular Weight: 126.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 119
Melting point (°C): 95

Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 119 °C
Melting Pt: 95 °C
MDL Number: MFCD00005523
CAS Number: 766-39-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5946f01f27d59655604a170e65df04953bfadeef&vid=5946f01f27d59655604a170e65df04953bfadeef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR