1-Chloro-3,5-dibromobenzene ≥96.0%
Supplier: TCI America
Synonyms:
3,5-Dibromochlorobenzene, 1,3-Dibromo-5-chlorobenzene
D3465-5G
D3465-25G
TCD3465-5GEA
50.56
CAD
TCD3465-5G
TCD3465-25G
1-Chloro-3,5-dibromobenzene ≥96.0%
1-Chloro-3,5-dibromobenzene
CAS Number: 14862-52-3
MDL Number: MFCD00070765
Molecular Formula: C6H3Br2Cl
Molecular Weight: 270.35
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 256
Melting point (°C): 96
Formula:
C₆H₃Br₂Cl MW: 270.35 g/mol Boiling Pt: 256 °C Melting Pt: 96 °C |
MDL Number:
MFCD00070765 CAS Number: 14862-52-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...