(3,5-Difluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D3462-5G
D3462-1G
TCD3462-5GEA
121.66
CAD
TCD3462-5G
TCD3462-1G
(3,5-Difluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(3,5-Difluorophenyl)acetic acid
CAS Number: 105184-38-1
MDL Number: MFCD00010316
Molecular Formula: C8H6F2O2
Molecular Weight: 172.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 69
Formula:
(C₆H₃F₂)CH₂COOH Melting Pt: 69 °C |
MDL Number:
MFCD00010316 CAS Number: 105184-38-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...