2,5-Dimethylbenzoxazole ≥98.0%

Supplier: TCI America

D3461-5G D3461-25G
TCD3461-5GEA 115.34 CAD
TCD3461-5G TCD3461-25G
2,5-Dimethylbenzoxazole ≥98.0%
2,5-Dimethylbenzoxazole

CAS Number: 5676-58-4
MDL Number: MFCD00005773
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 215
Flash Point (°C): 91
Specific Gravity (20/20): 1.09

Formula: C₉H₉NO
Boiling Pt: 215 °C
Density: 1.09 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00005773
CAS Number: 5676-58-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5166e0dc96388a63c52e02b5cefdee71e91c95f&vid=c5166e0dc96388a63c52e02b5cefdee71e91c95f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR