1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
1,3-Dimesityl-1H-imidazol-3-ium chloride, 1,3-Dimesitylimidazolium chloride
CAS Number: 141556-45-8
MDL Number: MFCD02684541
Molecular Formula: C21H25ClN2
Molecular Weight: 340.90
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 352
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: 1,3-Bis(2,4,6-trimethylphenyl)imidazolium Chloride, IMes.HCl
Formula:
C₂₁H₂₅ClN₂ MW: 340.89 g/mol Melting Pt: 352 °C |
MDL Number:
MFCD02684541 CAS Number: 141556-45-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...