2,5-Difluorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
D3436-25G
D3436-5G
TCD3436-25GEA
319.16
CAD
TCD3436-25G
TCD3436-5G
2,5-Difluorophenylboronic acid (contains varying amounts of Anhydride)
2,5-Difluorophenylboronic acid
CAS Number: 193353-34-3
MDL Number: MFCD01863171
Molecular Formula: C6H5BF2O2
Molecular Weight: 157.91
Form: Crystal
Color: White
Melting point (°C): 110
Other Product Names / Synonyms: 2,5-Difluorobenzeneboronic Acid
Formula:
C₆H₅BF₂O₂ Melting Pt: 110 °C |
MDL Number:
MFCD01863171 CAS Number: 193353-34-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...