2,3-Dichlorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

D3434-25G D3434-5G
TCD3434-25GEA 327.06 CAD
TCD3434-25G TCD3434-5G
2,3-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
2,3-Dichlorophenylboronic acid

CAS Number: 151169-74-3
MDL Number: MFCD01075703
Molecular Formula: C6H5BCl2O2
Molecular Weight: 190.81
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 125


Other Product Names / Synonyms: 2,3-Dichlorobenzeneboronic Acid

Formula: C₆H₅BCl₂O₂
Melting Pt: 125 °C
MDL Number: MFCD01075703
CAS Number: 151169-74-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1762e8a8b1c0cf5b533ce3ec29993a246f97c6a5&vid=1762e8a8b1c0cf5b533ce3ec29993a246f97c6a5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR