3',5'-Difluoroacetophenone ≥97.0%
Supplier: TCI America
Synonyms:
1-(3,5-Difluorophenyl)ethan-1-one
D3425-25G
D3425-5G
TCD3425-25GEA
210.14
CAD
TCD3425-25G
TCD3425-5G
3',5'-Difluoroacetophenone ≥97.0%
3',5'-Difluoroacetophenone
CAS Number: 123577-99-1
MDL Number: MFCD00042489
Molecular Formula: C8H6F2O
Molecular Weight: 156.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 95
Melting point (°C): 40
Flash Point (°C): 82
Formula:
C₈H₆F₂O MW: 156.13 g/mol Boiling Pt: 95 °C Melting Pt: 40 °C Flash Pt: 82 °C |
MDL Number:
MFCD00042489 CAS Number: 123577-99-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...