Difurfuryl sulfide ≥98.0%

Supplier: TCI America

D3420-25G D3420-5G
TCD3420-25GEA 183.44 CAD
TCD3420-25G TCD3420-5G
Difurfuryl sulfide ≥98.0%
Difurfuryl sulphide

CAS Number: 13678-67-6
MDL Number: MFCD00010081
Molecular Formula: C10H10O2S
Molecular Weight: 194.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 143
Flash Point (°C): 113
Freezing point (°C): 32


Other Product Names / Synonyms: Furfuryl Sulfide, 2,2'-(Thiodimethylene)difuran

Formula: C₁₀H₁₀O₂S
Boiling Pt: 143 °C
Flash Pt: 113 °C
MDL Number: MFCD00010081
CAS Number: 13678-67-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5c5b0afbeaf808b174ce5d545939a590fd6f5cc&vid=c5c5b0afbeaf808b174ce5d545939a590fd6f5cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR