6,11-Dihydrodibenzo[b,e]thiepin-11-one ≥98.0%

Supplier: TCI America

D3416-25G D3416-5G
TCD3416-25GEA 147.1 CAD
TCD3416-25G TCD3416-5G
6,11-Dihydrodibenzo[b,e]thiepin-11-one ≥98.0%
6,11-Dihydrodibenzo[b,e]thiepin-11-one

CAS Number: 1531-77-7
MDL Number: MFCD02093738
Molecular Formula: C14H10OS
Molecular Weight: 226.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 87

Formula: C₁₄H₁₀OS
MW: 226.29 g/mol
Boiling Pt: 180 °C
Melting Pt: 87 °C
MDL Number: MFCD02093738
CAS Number: 1531-77-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6483ab8b7987ae9a9ccd5936af86fde8433bab6d&vid=6483ab8b7987ae9a9ccd5936af86fde8433bab6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR