1,3-Di-tert-butyltoluene ≥98.0%
Supplier: TCI America
D3414-25G
D3414-5G
TCD3414-25GEA
110.76
CAD
TCD3414-25G
TCD3414-5G
1,3-Di-tert-butyltoluene ≥98.0%
1,3-Di-tert-butyltoluene
CAS Number: 15181-11-0
MDL Number: MFCD00026300
Molecular Formula: C15H24
Molecular Weight: 204.36
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 244
Flash Point (°C): 91
Freezing point (°C): 30
Other Product Names / Synonyms: 3,5-Di-tert-butyl-1-methylbenzene
Formula:
C₁₅H₂₄ Boiling Pt: 244 °C Flash Pt: 91 °C |
MDL Number:
MFCD00026300 CAS Number: 15181-11-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...