3,5-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
3,5-Dimethylbenzeneboronic acid, Dihydroxy(3,5-dimethylphenyl)borane, (3,5-Dimethylphenyl)metaboric acid
D3396-5G
D3396-25G
TCD3396-5GEA
72.68
CAD
TCD3396-5G
TCD3396-25G
3,5-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
3,5-Dimethylphenylboronic acid
CAS Number: 172975-69-8
MDL Number: MFCD00185689
Molecular Formula: C8H11BO2
Molecular Weight: 149.98
Form: Crystal
Color: White
Melting point (°C): 237
Other Product Names / Synonyms: 3,5-Dimethylbenzeneboronic Acid, m-Xylene-5-boronic Acid
Formula:
C₈H₁₁BO₂ MW: 149.98 g/mol Melting Pt: 237 °C |
MDL Number:
MFCD00185689 CAS Number: 172975-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...