2,4-Difluorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
D3391-25G
D3391-5G
TCD3391-25GEA
216.46
CAD
TCD3391-25G
TCD3391-5G
2,4-Difluorophenylboronic acid (contains varying amounts of Anhydride)
2,4-Difluorophenylboronic acid
CAS Number: 144025-03-6
MDL Number: MFCD01318998
Molecular Formula: C6H5BF2O2
Molecular Weight: 157.91
Form: Crystal
Color: White
Melting point (°C): 250
Other Product Names / Synonyms: 2,4-Difluorobenzeneboronic Acid
Formula:
C₆H₅BF₂O₂ Melting Pt: 250 °C |
MDL Number:
MFCD01318998 CAS Number: 144025-03-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...