4,4''-Diamino-p-terphenyl ≥98.0%

Supplier: TCI America

D3390-1G D3390-5G
TCD3390-1GEA 150.1 CAD
TCD3390-1G TCD3390-5G
4,4''-Diamino-p-terphenyl ≥98.0%
4,4''-Diamino-p-terphenyl

CAS Number: 3365-85-3
MDL Number: MFCD00051532
Molecular Formula: C18H16N2
Molecular Weight: 260.34
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 242

Formula: C₁₈H₁₆N₂
MW: 260.34 g/mol
Melting Pt: 242 °C
MDL Number: MFCD00051532
CAS Number: 3365-85-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f161894d06c9e0b740940d75c7ec3989d332e7&vid=16f161894d06c9e0b740940d75c7ec3989d332e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR